ContactUsLinkComponent
FLUORESCENT DYE 594-I PHALLOIDIN FLUORESCENCE DYE 594-I WITH PHALLOIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594- I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE OF 1 * 300 Essai
Référence ABNOU0292
Fournisseur:  Abnova
FLUORESCENT DYE 594-I PHALLOIDIN FLUORESCENCE DYE 594-I WITH PHALLOIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594- I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE OF 1 * 300 Essai
Référence ABNOU0292
Fournisseur:  Abnova
Informations associées
Frequently Bought Together