ContactUsLinkComponent
FLUORESCENT DYE 594-I STREPTAVIDIN FLUORESCENCE DYE 594-I WITH STREPTAVIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594-I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE 1 * 200 µG
Référence ABNOU0291
Fournisseur:  Abnova
FLUORESCENT DYE 594-I STREPTAVIDIN FLUORESCENCE DYE 594-I WITH STREPTAVIDIN CONJUGATE. BIOMOLECULES CONJUGATED TO FLUORESCENCE DYE 594-I EXHIBIT LITTLE SPECTRAL OVERLAP WITH GREEN-FLUORESCENT CONJUGATES, AND CAN BE EFFICIENTLY EXCITED BY 568NM LINE 1 * 200 µG
Référence ABNOU0291
Fournisseur:  Abnova
Informations associées
Frequently Bought Together