ContactUsLinkComponent
KAT5 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
N° de catalogue ABNOH00010524-PW2
Fournisseur:  Abnova
KAT5 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
N° de catalogue ABNOH00010524-PW2
Fournisseur:  Abnova
Frequently Bought Together