Trypsin protease, MS grade
Trypsin Protease, frozen, 1×100 µg, 100 µG
N° de catalogue PIER90305
Fournisseur: Thermo Fisher Scientific
Trypsin protease, MS grade
N° de catalogue PIER90305
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Aller à
Product Details & Documents
Pierce™ Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
- Cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
- No detectable chymotrypsin activity thus making it highly pure
- Chemically modified for reduced autolytic activity
Applications include in-gel digestion of proteins from 1-D or 2-D gels and in-solution tryptic digestion of proteins. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. It has been treated with TPCK to eliminate chymotryptic activity, improving digestion specificity. The enzyme is most active in pH ranges 7 to 9 and can be reversibly inactivated at pH <4.
Documents
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- Cdt:100 µG
- Désignation:Trypsin Protease, frozen, 1×100 µg