Mouse serum
Mouse serum
Sérum de souris, Filtré de manière stérile, Frozen
N° de catalogue S2160-010
Fournisseur:   Biowest
Mouse serum
N° de catalogue S2160-010
Fournisseur:  Biowest
Mouse serum
Mouse serum
Sérum de souris, Filtré de manière stérile, Frozen
Prix
52.60 SFr.
Connexion Pour la disponibilité
1 * 10 mL
Product Details & Documents
Mouse serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Mouse serum is used as a blocking agent in immunoassays and controls.
Spécifications
  • Format du sérum:
    Frozen
  • Stérilité:
    Filtré de manière stérile
  • Désignation:
    Sérum de souris