Rabbit serum
Sérum de lapin, inactivé par la chaleur, Filtré de manière stérile, Frozen
N° de catalogue S250H-500
Fournisseur: Biowest
Aller à
Product Details & Documents
Rabbit serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Rabbit serum is used as a blocking agent in immunoassays and controls.
Spécifications
- Format du sérum:Frozen
- Stérilité:Filtré de manière stérile
- Désignation:Sérum de lapin, inactivé par la chaleur
- Seq:52