Human recombinant BIN2 (from E. coli)
50 µG
N° de catalogue BIRBORB49226-50
Marketsource
Fournisseur: Biorbyt
Human recombinant BIN2 (from <i>E. coli</i>)
N° de catalogue BIRBORB49226-50
Fournisseur: Biorbyt
Marketsource
Numéro CAS:
Aller à
Product Details & Documents
Documents
orb49226
Fiche technique
Spécifications
Spécifications
- Cdt:50 µG
- Purity:>95% as determined by reducing SDS-PAGE.
- Synonyme de protéine:Bridging Integrator 2
- Conjugaison:Unconjugated
- Base de données UnitProt:Q9UBW5 (Human)
- Sequence:MGSSHHHHHHSSGLVPRGSHMAEGKAGGAAGLFAKQVQKKFSRAQEKVLQKLGKAVETKDERFEQ SASNFYQQQAEGHKLYKDLKNFLSAVKVMHESSKRVSETLQEIYSSEWDGHEELKAIVWNNDLLW EDYEEKLADQAVRTMEIYVAQFSEIKERIAKRGRKLVDYDSARHHLEAVQNAKKKDEAKTAKAEE EFNKAQTVFEDLNQELLEELPILYNSRIGCYVTIFQNISNLRDVFYREMSKLNHNLYEVMSKLEK QHSN
- Source:E. coli
- Cat. No.:BIRBORB49226-50
- Conditions de stockage:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
- Tested applications:SDS-PAGE
- Species:Human
- Nom de la protéine/du peptide:BIN2
- Type de protéine/peptide:Recombinant
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



