Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Rat recombinant IL2

Rat recombinant IL2

5 µG
Catalog # BIRBORB80415-5
Marketsource
Supplier:   Biorbyt
Rat recombinant IL2
Catalog # BIRBORB80415-5
Supplier:  Biorbyt
Marketsource
CAS Number:  
Rat recombinant IL2

5 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 5 µG
Product Details & Documents

Documents

Specifications

Specifications

  • Cdt:
    5 µG
  • Activité biologique:
    The ED50 as determined by the dose-dependent stimulation of the proliferation of Mouse CTLL-2 cells is < 0.4 ng/ml, corresponding to a specific activity of > 2.5 x 106 units/mg.
  • Purity:
    >98%
  • Synonyme de protéine:
    TCGF|T-cell growth factor
  • Base de données UnitProt:
    P17108
  • Sequence:
    MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAIAQSI ISTM
  • Cat. No.:
    BIRBORB80415-5
  • Conditions de stockage:
    Store at −20 °C.
  • Formulation:
    The protein (1mg/ml) was lyophilized without any additives.
  • Solubilité:
    It is recommended to reconstitute the lyophilized Interleukin in sterile 100mM AcOH not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Tested applications:
    SDS-PAGE
  • Species:
    Rat
  • Nom de la protéine/du peptide:
    IL2
  • Type de protéine/peptide:
    Recombinant

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.