Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Sheep leptin (from<i> E. coli</i>)

Sheep leptin (from E. coli)

100 µG
N° de catalogue BIRBORB80492-100
Marketsource
Fournisseur:   Biorbyt
Sheep leptin (from<i> E. coli</i>)
N° de catalogue BIRBORB80492-100
Fournisseur:  Biorbyt
Marketsource
Numéro CAS:  
Sheep leptin (from E. coli)

100 µG

Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 100 µG
Product Details & Documents

Documents

Spécifications

Spécifications

  • Cdt:
    100 µG
  • Cat. No.:
    BIRBORB80492-100
  • Conditions de stockage:
    Lyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution with 0.0045mM NaHCO3.
  • Activité biologique:
    Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
  • Solubilité:
    It is recommended to reconstitute the lyophilized Leptin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Synonyme de protéine:
    LEP
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Val-Pro-Ile-Arg.
  • Tested applications:
    SDS-PAGE
  • Species:
    Sheep
  • Nom de la protéine/du peptide:
    Leptin
  • Source:
    E. coli

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.