Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Sheep leptin (from<i> E. coli</i>)

Sheep leptin (from E. coli)

1 mg
N° de catalogue BIRBORB80492-1
Marketsource
Fournisseur:   Biorbyt
Sheep leptin (from<i> E. coli</i>)
N° de catalogue BIRBORB80492-1
Fournisseur:  Biorbyt
Marketsource
Numéro CAS:  
Sheep leptin (from E. coli)

1 mg

Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 mg
Product Details & Documents

Documents

Spécifications

Spécifications

  • Cdt:
    1 mg
  • Cat. No.:
    BIRBORB80492-1
  • Conditions de stockage:
    Lyophilized Leptin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Leptin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution with 0.0045mM NaHCO3.
  • Activité biologique:
    Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
  • Solubilité:
    It is recommended to reconstitute the lyophilized Leptin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Synonyme de protéine:
    LEP
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Ala-Val-Pro-Ile-Arg.
  • Tested applications:
    SDS-PAGE
  • Species:
    Sheep
  • Nom de la protéine/du peptide:
    Leptin
  • Source:
    E. coli

Spécifications

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..