L-Phénylalanine 98.5+%
Flacon plastique, 500 g
: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
- :63-91-2
- :C₉H₁₁NO₂
- :MFCD00064227
- :165.19 g/mol
- :14,07271
- :313 °C (1013 hPa)
- :270…275 °C (déc.)
- :0,754 g/cm³ (20 °C)
- :Température ambiante
- :200-568-1
Documents
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- pH:5.4 to 6.0 (1 % in water)
- Specific optical rotation:-33.0° to -35.2° (20°C, 589 nm) (c=2, water) (on dried substance)
- Titration with HClO4:98.50 to 102.00 % (On dried basis)
- Lead (Pb):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
- Type de conditionnement:Flacon plastique
- Cdt:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



