Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Laboratory coats, PP, with snap closure

With snap closure, XL
N° de catalogue HOIT114526
Laboratory coats, PP, with snap closure
N° de catalogue HOIT114526
Laboratory coats, PP, with snap closure
Laboratory coats, PP, with snap closure

With snap closure, XL

Prix
SFr. 144.00
Connexion Pour la disponibilité
1 * 50 ST
Product Details & Documents

These disposable laboratory coats made from PP material.

  • With shirt, Korean, knitted collar
  • Snaps for closures

Weight: 25 to 55 g/m²

Colour: White.

Packaging: 5 pieces/ bag, 10 bags/case.

Spécifications
  • Taille de confection:
    XL
  • Désignation:
    With snap closure
  • Seq:
    02
  • Cat. No.:
    HOIT114526
  • Poids:
    25 - 55 g/m²
  • Applications des tenues:
    Laboratoire
  • Genre:
    Unisex
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.