Heating tapes, HST
Heating tape HST with 0,5 m length, 30 W output
N° de catalogue HORS025201
Fournisseur: HORST
Aller à
Product Details & Documents
These slim heating tapes consist of two PTFE insulated heating conductors with earthed protection braid which have a common coating made of metal cloth.
Spécifications
- Désignation:Heating tape HST with 0,5 m length, 30 W output
- Seq:01