Inserts
E 484 1/1 insert, wire mesh for various utensils, 8 mm mesh spacing, G 7823, G 7824, G 7825, G 7826, PG 8528, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 150×470×480 mm
MIEL5109190
- Désignation:E 484 1/1 insert, wire mesh for various utensils, 8 mm mesh spacing
- Seq:18
- l×p×h:150×470×480 mm
- Pour:G 7823, G 7824, G 7825, G 7826, PG 8528, PG 8535/36, PG 8582, PG 8582 CD, PG 8592