Filling solutions for combined/reference electrodes
3M KCl solution, saturated with AgCl, for appropriate Ag/AgCl (pHC3xxx/REF3xx) reference elements, 100 ml bottle, 100 mL
N° de catalogue HACHS21M004
Fournisseur: Hach
Filling solutions for combined/reference electrodes
N° de catalogue HACHS21M004
Fournisseur: Hach
Numéro CAS:
Aller à
Product Details & Documents
Filling and storage solutions for combined and reference electrodes.
Certificats
Rechercher par numéro de lotSpécifications
Spécifications
- Cdt:100 mL
- Désignation:3M KCl solution, saturated with AgCl, for appropriate Ag/AgCl (pHC3xxx/REF3xx) reference elements, 100 ml bottle