ContactUsLinkComponent
THE EXOMIR™-MINI KIT IS DESIGNED FOR FILTRATION-BASED CAPTURE OF EXOSOMES AND OTHER MICROPARTICLES FROM SMALL VOLUMES OF CELL-FREE B IOFLUIDS, AND FOR EXTRACTION OF RNA FROM THE CAPTURED PARTICLES. 1 * 1 KIT
Référence BIOO5148-01
Fournisseur:  BIOO SCIENTIFIC
THE EXOMIR™-MINI KIT IS DESIGNED FOR FILTRATION-BASED CAPTURE OF EXOSOMES AND OTHER MICROPARTICLES FROM SMALL VOLUMES OF CELL-FREE B IOFLUIDS, AND FOR EXTRACTION OF RNA FROM THE CAPTURED PARTICLES. 1 * 1 KIT
Référence BIOO5148-01
Fournisseur:  BIOO SCIENTIFIC
Frequently Bought Together