THE PROPRIETARY NEUROAIM™ TRANSDUCTION REAGENT IS USED FOR THE DIRECTED DELIVERY OF RNAI AGENTS INTO NEURONAL CELLS (BOTH IN PRIMARY CELLS AND IN CELL LINES). 1 * 0,4 mL
Référence BIOO515201
Fournisseur: BIOO SCIENTIFIC
Frequently Bought Together