96-Well Blocks
96-well block with cover foils, 2 blocks/foils, 2 ml
Catalog # ZYMOP1002-2
Marketsource
Supplier: Zymo Research
Jump to:
Product Details & Documents
Deep-well block, 2 ml for processing and mixing of samples.
- Conforms to laboratory standards
96-well block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2 ml per round bottom well.
Specifications
- Emballage:2 blocks/foils
- Désignation:96-well block with cover foils
- Seq:02
- Volume total:2 ml