Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Quick-DNA™ Kits

Quick-DNA™ Kits

Quick-DNA™ 96 kit, 10×96 preps, 1 KIT
N° de catalogue ZYMOD3012
Marketsource
Fournisseur:   Zymo Research
Quick-DNA™ Kits
N° de catalogue ZYMOD3012
Fournisseur:  Zymo Research
Marketsource
Numéro CAS:  
Quick-DNA™ Kits
Quick-DNA™ Kits

Quick-DNA™ 96 kit, 10×96 preps, 1 KIT

Numéro CAS:  
Prix
SFr. 1’311.00
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents

DNA from cells, swabs, and whole blood.

  • Quick & easy: Simple 20 minute procedure
  • Unique technology excludes the use of proteinase K and organic denaturants
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more

The Quick-DNA™ kits are ideal for easy, rapid isolation of total DNA (e.g., genomic, mitochondrial, viral) from a variety of biological sample sources. Whole blood (fresh or stored), buffy coat, buccal cells, cells from culture, saliva, and other biological liquid samples can be processed with these kits. Zymo-Spin™ column/plate technology enables high-quality DNA purification in minutes. PCR inhibitors are effectively removed, and the eluted DNA is ideal for PCR, nucleotide blotting, DNA sequencing, restriction endonuclease digestion, bisulfite conversion/methylation analysis, and other downstream applications.

Spécifications

Spécifications

  • Cdt:
    1 KIT
  • Désignation:
    Quick-DNA™ 96 kit, 10×96 preps
  • Cat. No.:
    ZYMOD3012
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Multiwell plate
  • Sample type:
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Capacité d'échantillons:
    5 µg
  • Désignation:
    Quick-DNA™ 96 kit
  • Yield:
    Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Taille de l'échantillon:
    ≤1×10⁶ cells or 5 mg tissue

Spécifications