Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Quick-DNA™ Kits

Quick-DNA™ Kits

Quick-DNA™ Microprep kit, 200 preps, 1 KIT
Catalog # ZYMOD3021
Marketsource
Supplier:   Zymo Research
Quick-DNA™ Kits
Catalog # ZYMOD3021
Supplier:  Zymo Research
Marketsource
CAS Number:  
Quick-DNA™ Kits
Quick-DNA™ Kits

Quick-DNA™ Microprep kit, 200 preps, 1 KIT

CAS Number:  
Price
SFr. 489.00
Log in for availability
1 * 1 KIT
Product Details & Documents

DNA from cells, swabs, and whole blood.

  • Quick & easy: Simple 20 minute procedure
  • Unique technology excludes the use of proteinase K and organic denaturants
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more

The Quick-DNA™ kits are ideal for easy, rapid isolation of total DNA (e.g., genomic, mitochondrial, viral) from a variety of biological sample sources. Whole blood (fresh or stored), buffy coat, buccal cells, cells from culture, saliva, and other biological liquid samples can be processed with these kits. Zymo-Spin™ column/plate technology enables high-quality DNA purification in minutes. PCR inhibitors are effectively removed, and the eluted DNA is ideal for PCR, nucleotide blotting, DNA sequencing, restriction endonuclease digestion, bisulfite conversion/methylation analysis, and other downstream applications.

Specifications

Specifications

  • Cdt:
    1 KIT
  • Désignation:
    Quick-DNA™ Microprep kit, 200 preps
  • Cat. No.:
    ZYMOD3021
  • Prep size:
    Micro
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Capacité d'échantillons:
    5 µg
  • Désignation:
    Quick-DNA™ microprep kit
  • Yield:
    Up to 5 µg/well total DNA is eluted into ≥30 µl DNA Elution Buffer or water. Human whole blood yields 3 - 7 µg DNA per 100 µl blood sampled. Mammalian tissues already homogenized will yield 1 - 3 µg DNA per mg.
  • Taille de l'échantillon:
    ≤1×10⁶ cells or 5 mg tissue

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.