Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Zymo-Spin™ Column/Filter Assemblies

Zymo-Spin™ Column/Filter Assemblies

Zymo-Spin™ VI with Reservoir, Spin column/filter assembly, 20
N° de catalogue ZYMOC1018-20
Marketsource
Fournisseur:   Zymo Research
Zymo-Spin™ Column/Filter Assemblies
N° de catalogue ZYMOC1018-20
Fournisseur:  Zymo Research
Marketsource
Numéro CAS:  
Zymo-Spin™ Column/Filter Assemblies
Zymo-Spin™ Column/Filter Assemblies

Zymo-Spin™ VI with Reservoir, Spin column/filter assembly, 20

Numéro CAS:  
Prix
121.00 SFr.
Connexion Pour la disponibilité
1 * 20 ST
Product Details & Documents

Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.

  • For purifying DNA and or/RNA from high-volume sample inputs
  • High-recovery

The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA.

The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.

The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥1 ml elution buffer or water. The capacity of the spin-column with filter is 75 ml.

The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1,6 μm pore size glass fibre filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.

Spécifications

Spécifications

  • Cdt:
    20
  • Désignation:
    Zymo-Spin™ VI with Reservoir
  • Format:
    Spin column/filter assembly

Spécifications