Quick-DNA™ Tissue/Insect Kits
Product Details & Documents
Simple and efficient isolation of DNA from insects, including mosquitoes, bees, lice, ticks and D. melanogaster.
- Also compatible with tough-to-lyse tissues from other organisms
- Ultra-high density BashingBeads™ are fracture resistant and chemically inert
The Quick-DNA™ Tissue/Insect Kits are designed for the simple and rapid isolation of DNA (e.g., genomic, viral, mitochondrial) from fresh, frozen, or stored insect specimens - including mosquitoes, bees, lice, ticks, and D. melanogaster. The procedures are easy and can be completed in minutes. All kits are compatible with mammalian tissues, whole blood, and cultured cells. Samples are rapidly and efficiently lysed by bead beating with our state-ofthe- art, ultra-high density BashingBeads™. The DNA is then isolated and purified using our Zymo-Spin™ technology and is ideal for downstream molecular-based applications including PCR, array, genotyping, etc.
Spécifications
Spécifications
- Cdt:2 KIT
- Désignation:Quick-DNA™ tissue/insect microprep kit, 2×96 preps
- Format:96 well spin column plate
- Cat. No.:ZYMOD6017
- Prep size:Mini
- Refine target molecule:Total DNA
- Target molecule:DNA
- Method / Format:Multiwell plate
- Sample type:Cultured cells|Invertebrates|Human/animal cells and tissues
- Désignation:Quick-DNA™ tissue/insect 96 kit
Spécifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



