Racks and boxes
Rack for cryotubes with star foot, —, 40, jaune, 219×109×26 mm
N° de catalogue TPPA99016
Fournisseur: TPP
Aller à
Product Details & Documents
These reusable cryotube racks and boxes with alphanumerical inscription are ideal for sample identification.
- Polypropylene, heat resistant, autoclavable material
- Racks can be quickly assembled and disassembled thus space saver
- Cryo-box, single handling of lid even after multiple use
- Cryo-rack, locking system in the groves allowing one hand operation when used with star-shape foot cryo tube
: ISO 9001:2008 certified.
Spécifications
- Pour tubes:—
- Nb de places:40
- Couleur:jaune
- Désignation:Rack for cryotubes with star foot
- Seq:05
- l×p×h:219×109×26 mm