Racks and boxes
Racks and boxes
Rack for cryotubes with star foot, —, 40, jaune, 219×109×26 mm
N° de catalogue TPPA99016
Fournisseur:   TPP
Racks and boxes
N° de catalogue TPPA99016
Fournisseur:  TPP
Racks and boxes
Racks and boxes
Rack for cryotubes with star foot, —, 40, jaune, 219×109×26 mm
Prix
SFr. 29.40
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These reusable cryotube racks and boxes with alphanumerical inscription are ideal for sample identification.
  • Polypropylene, heat resistant, autoclavable material
  • Racks can be quickly assembled and disassembled thus space saver
  • Cryo-box, single handling of lid even after multiple use
  • Cryo-rack, locking system in the groves allowing one hand operation when used with star-shape foot cryo tube
 : ISO 9001:2008 certified.
Spécifications
  • Pour tubes:
  • Nb de places:
    40
  • Couleur:
    jaune
  • Désignation:
    Rack for cryotubes with star foot
  • Seq:
    05
  • l×p×h:
    219×109×26 mm