Murine IL-4 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinan t murine IL-4 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and soluti 1 * 200 Tests
Référence PEPR900-M49
Fournisseur: PeproTech, Inc.
Frequently Bought Together