Rat VEGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant r at VEGF in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reagents a 1 * 200 Tests
Catalog # PEPR900-M436
Supplier: PeproTech, Inc.
Frequently Bought Together