Stoppers, for test tubes
naturel, 16 - 17 mm
N° de catalogue RATI3914900
Fournisseur: RATIOLAB
Aller à
Product Details & Documents
PE.
- Very tight seal due to two sealing rings
- Available in natural and four more colors
Spécifications
- Ø inférieur:16 - 17 mm
- Couleur:naturel
- Seq:05