Trypsin protease, MS grade
Trypsin protease, MS grade
Trypsin Protease, 5×100 µg, 1 KIT
N° de catalogue PIER90058
Fournisseur:   Thermo Fisher Scientific
Trypsin protease, MS grade
N° de catalogue PIER90058
Numéro CAS:  
Trypsin protease, MS grade
Trypsin protease, MS grade
Trypsin Protease, 5×100 µg, 1 KIT
Numéro CAS:  
Prix
665.00 SFr.
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents
Pierce™ Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
  • Cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
  • No detectable chymotrypsin activity thus making it highly pure
  • Chemically modified for reduced autolytic activity
Applications include in-gel digestion of proteins from 1-D or 2-D gels and in-solution tryptic digestion of proteins. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. It has been treated with TPCK to eliminate chymotryptic activity, improving digestion specificity. The enzyme is most active in pH ranges 7 to 9 and can be reversibly inactivated at pH <4.
Documents
Spécifications
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    Trypsin Protease, 5×100 µg