Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

2-Méthylthiazole

Glass bottle for solids, 25 g
Catalog # ACTIAS23101G25
Marketsource
Supplier:   ACTIVATE SCIENTIFIC
2-Méthylthiazole
Catalog # ACTIAS23101G25
Marketsource
CAS Number:  3581-87-1
2-Méthylthiazole

Glass bottle for solids, 25 g

CAS Number:  3581-87-1
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:3581-87-1
  • Formula:C₄H₅NS
  • MDL Number:MFCD00053144
  • Molecular Weight:99.16 g/mol
  • EINECS:222-702-8
Specifications

Specifications

  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    25 g

Specifications