Thymidine
Glass bottle for solids, 25 g
  ACTIAS21759G25
Marketsource
Thymidine
  ACTIAS21759G25
Marketsource
 :  50-89-5
Thymidine
Glass bottle for solids, 25 g
 :  50-89-5
1 * 25 g
 :  1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine
  •  :50-89-5
  •  :C₁₀H₁₄N₂O₅
  •  :MFCD00006537
  •  :242.23 g/mol
  •  :Température ambiante
  •  :200-070-4
  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    25 g