Vert de bromocrésol ACS
Glass bottle for solids, 5 g
CAS No.: 76-60-8 Molecular Formula: C21H14Br4O5S Formula Weight: 698.04 Physical Form: Powder Appearance: Pale brown to yellow Melting Point: 225° dec. Beilstein Reference: 372527 Merck Reference: 14,1386 MDL No.: MFCD00005874
: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- :76-60-8
- :C₂₁H₁₄Br₄O₅S
- :MFCD00005874
- :698.02 g/mol
- :14,1386
- :225 °C (déc.)
- :Température ambiante
- :200-972-8
- Type de conditionnement:Glass bottle for solids
- Cdt:5 g