Buffers, sodium phosphate, 0,5 M solutions
Sodium phosphate, buffer solution, 0,5 M, 7,5, Flacon Verre, 250 mL
Fournisseur: Thermo Fisher Scientific
Buffers, sodium phosphate, 0,5 M solutions
N° de catalogue J61561.AK
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Aller à
Product Details & Documents
Physical form: liquid
MDL No.: MFCD00146206
Documents
Spécifications
Spécifications
- Concentration:0,5 M
- Type de conditionnement:Flacon Verre
- Cdt:250 mL
- Désignation:Sodium phosphate, buffer solution
- Valeur pH:7,5
- Cat. No.:J61561.AK
- Physical form:Liquid
- MDL No.:MFCD00146206
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



