Safety helmets, Concept HeightMaster
Safety helmet, vented, without ratchet, EU, jaune
N° de catalogue CENUS08EYFM
Fournisseur: CENTURION SAFETY PRODUCTS
Safety helmets, Concept HeightMaster
N° de catalogue CENUS08EYFM
Fournisseur: CENTURION SAFETY PRODUCTS
Aller à
Product Details & Documents
Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene). The Concept HeightMaster has been designed for applications where the wearer is working at height, where the suspension system and the helmet design and structure will reduce the risk of the helmet coming off as a result of an impact or a fall.
- With extended nape area
- Six point terylene inner cradle
- Highly absorbent hydro-flock sweatband
- With increased side rigidity (additional lateral deformity test)
- Ventilation through wide ventilation slots
- With or without ratchet, with full peak
Size: 510 to 630 mm
Weight: 325 g
Certifications: EN 397, EN 12492, EN 50365
Spécifications
- Couleur:jaune
- Désignation:Safety helmet, vented, without ratchet, EU
- Seq:06
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- CENUS30VPFermetures des orfices de ventilation pour casques ventilés ConceptConnexion Pour la disponibilité1 * 1 ST
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
- CENUS31PGarniture frontale: S31P, PVC, pour casques ConceptConnexion Pour la disponibilité1 * 1 ST - SFr. 2.20