Sample containers, straight, with screw cap, WHEATON®
100 ml, Printed label, sterile, incolore, PS/Métal
N° de catalogue 216-1014
Fournisseur: DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
N° de catalogue 216-1014
Fournisseur: DWK Life Sciences
Aller à
Product Details & Documents
Polystyrene clear, with metal screw cap.
- Manufactured from FDA approved virgin polystyrene
- Supplied sterile and for in vitro use only
- CE marked in accordance with European Directive 98/79/EC
- All containers are leak tested to British Standard BS5213:1975(1989)
- All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Caution: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904
Spécifications
- Type:Printed label, sterile
- Capacité:100 ml
- Couleur:incolore
- Matière:PS/Métal