Order Entry
Switzerland
ContactUsLinkComponent
[EN]GAS25H2S 100CO 2.2%CH4 18%O2 N2 110L 1 * 1 Pce
Catalog # BWTE4GA18X22W25H10
Supplier:  BW TECHNOLOGIES
[EN]GAS25H2S 100CO 2.2%CH4 18%O2 N2 110L 1 * 1 Pce
Catalog # BWTE4GA18X22W25H10
Supplier:  BW TECHNOLOGIES
Frequently Bought Together