Lumefantrine, Reference Standards
Lumefantrine, Reference Standards
Lumefantrine, USP reference standard, Vial, 100 mg
N° de catalogue 1370746.
Fournisseur:   USP
Lumefantrine, Reference Standards
N° de catalogue 1370746.
Fournisseur:  USP
Numéro CAS:  
Lumefantrine, Reference Standards
Lumefantrine, USP reference standard, Vial, 100 mg
Numéro CAS:  
Prix
SFr. 470.00
Connexion Pour la disponibilité
1 * 100 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Spécifications
Spécifications
  • Type de conditionnement:
    Vial
  • Cdt:
    100 mg
  • Normes:
    USP reference standard
  • Désignation:
    Lumefantrine
  • Cat. No.:
    1370746.
  • N ° CAS.:
    82186-77-4
  • Désignation:
    Lumefantrine
Spécifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..