Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Zonisamide, Reference Standards

Zonisamide Related Compound C, USP reference standard, Vial, 20 mg
N° de catalogue 1725036.
Fournisseur:   USP
Zonisamide, Reference Standards
N° de catalogue 1725036.
Fournisseur:  USP
Numéro CAS:  
Zonisamide, Reference Standards

Zonisamide Related Compound C, USP reference standard, Vial, 20 mg

Numéro CAS:  
Prix
SFr. 1’360.00
Connexion Pour la disponibilité
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Spécifications

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    20 mg
  • Normes:
    USP reference standard
  • Désignation:
    Zonisamide Related Compound C
  • Cat. No.:
    1725036.
  • Désignation:
    Zonisamide Related Compound C
  • Synonyme(s):
    N'-(benzisoxazol-3-ylmethylsulfonyl)-N,N-dimethylformimidamide

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.