Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Zonisamide, Reference Standards

Zonisamide, Reference Standards

Zonisamide Related Compound A, USP reference standard, Vial, 20 mg
Catalog # 1725014.
Supplier:   USP
Zonisamide, Reference Standards
Catalog # 1725014.
Supplier:  USP
CAS Number:  
Zonisamide, Reference Standards

Zonisamide Related Compound A, USP reference standard, Vial, 20 mg

CAS Number:  
Price
SFr. 1’440.00
Log in for availability
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Type de conditionnement:
    Vial
  • Cdt:
    20 mg
  • Normes:
    USP reference standard
  • Désignation:
    Zonisamide Related Compound A
  • Cat. No.:
    1725014.
  • N ° CAS.:
    73101-64-1
  • Désignation:
    Zonisamide Related Compound A
  • Synonyme(s):
    1,2-benzisoxazole-3-methanesulfonic acid sodium salt

Specifications