8 COMPOSES Fluorides (F-) 10mg/l Bromides (Br-) 10mg/l Nitrites (NO2-) 10mg/l Phosphates (PO43-) 100mg/l Sulphates (SO42-) 1000mg/l Chlorides (Cl-) 500mg/l Nitrates (NO3-) 500mg/l Bromates (BrO3-) 1mg/l DANS H2O 1 * 500 mL
N° de catalogue 89226.260
Fournisseur: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together