Scratching tools, Machu
Scratching tool, Basic
N° de catalogue SIMEVF8600
Fournisseur: Industrial Physics
Aller à
Product Details & Documents
These scratching tools can perform a Machu corrosion test. Each tool is provided with a 1 mm width cutter to cut the coating down to the metal.
- Ergonomic shaped grip
- Easy to replace the cutter
- Mandatory test in Qualicoat and QIB accredited laboratories
CC2000 has a self positioning knife bracket to expose the substrate with a perpendicular cut through the coating. CC3000 has an adjustable cutting depth and two ball bearings to guide the cut and this guarantees reproducible results.
Certifications: ISO 17872. DIN 53167, 50021, ISO 4628-8, 9227.
Documents
TQC Company Video
Vidéos
Spécifications
- Désignation:Scratching tool, Basic