Adapters, expansion
, Expansion, 34/35 NS, 45/40 NS, ,
N° de catalogue WITG0.510.019
Fournisseur: witeg Labortechnik
Aller à
Product Details & Documents
Borosilicate glass 3.3
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapters.
Certifications: According to DIN 12257.
Spécifications
- Rodage mâle:34/35 NS
- Rodage femelle:45/40 NS
- Type:Expansion
- Seq:03