ContactUsLinkComponent
elproLOG USER base licence User Management software V1.00.12 User Management for all elproLOG software components. System requireme nts: elproLOG USER can only be used together with elproLOG software components version 3.6 or newer null
N° de catalogue ELPR800386
Fournisseur:  ELPRO-BUCHS
elproLOG USER base licence User Management software V1.00.12 User Management for all elproLOG software components. System requireme nts: elproLOG USER can only be used together with elproLOG software components version 3.6 or newer null
N° de catalogue ELPR800386
Fournisseur:  ELPRO-BUCHS
Frequently Bought Together