Buffer, TBS solution 10X concentrate (TRIS buffered saline) with phenol red, pH 7,4
TBS buffer solution 10X concentrate with phenol red, pH 7,4, 250 ml
N° de catalogue CAYM600220-250
Fournisseur: Cayman Chemical
Buffer, TBS solution 10X concentrate (TRIS buffered saline) with phenol red, pH 7,4
N° de catalogue CAYM600220-250
Fournisseur: Cayman Chemical
Numéro CAS:
Aller à
Product Details & Documents
250 mM Tris-base, pH 7,4, 1,37 M sodium chloride, containing 27 mM KCl, 0,4 mM phenol red.
Suggested storage (long term) temperature: 4 °C.
Spécifications
Spécifications
- Cdt:250 ml
- Désignation:TBS buffer solution 10X concentrate with phenol red, pH 7,4



