PPM-18 ≥95%
Glass vial, 5 mg
N° de catalogue CAYM13327-5
Fournisseur: Cayman Chemical
Aller à
Product Details & Documents
- Inhibits activation of NF-κB
An inhibitor NF-κB activation in vitro and in vivo (IC₅₀= 5 µm).
Synonymes: NSC 73233
- Numéro CAS:65240-86-0
- Température de stockage:Congélateur
Spécifications
Résultats des tests de spécification
- Formulation:A crystalline solid
- Purity:≥95%
- Stability:≥2 years
Spécifications
- Type de conditionnement:Glass vial
- Cdt:5 mg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



