GN25
5 mg
Katalognr. CAYM10948-5
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
A 2-thio-dimethoxy naphthoquinone analog that blocks Snail binding to p53 and induces p53 expression in cancer cells in a K-Ras dependent manner
Synonyme: 3-[(1,4-dihydro-5,8-dimethoxy-1,4-dioxo-2-naphthalenyl)thio]-propanoic acid, p53-Snail binding Inhibitor
- CAS Nummer:1227401-27-5
- Formel:C₁₅H₁₄O₆S
- Molekulargewicht:322.3 g/mol
Technische Daten
Technische Daten
- Cdt:5 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



