Veterinary needles
Veterinary needles
Veterinary Luer needles, 18, 25 mm
N° de catalogue POPR7212
Fournisseur:   CADENCE SCIENCE
Veterinary needles
N° de catalogue POPR7212
Fournisseur:  CADENCE SCIENCE
Veterinary needles
Veterinary needles
Veterinary Luer needles, 18, 25 mm
Prix
1 810.00 SFr.
Connexion Pour la disponibilité
1 * 12 ST
Product Details & Documents
These hypodermic needles are all fitted with a large knurled nickel plated hub which allows for secure gripping even under the most difficult circumstances.
Spécifications
  • Jauge:
    18
  • Désignation:
    Veterinary Luer needles
  • Longueur:
    25 mm
  • Seq:
    01