Dextran sulphate
Bottle
N° de catalogue RC-043
Fournisseur: G-Biosciences
Aller à
Product Details & Documents
Dextran sulfate sodium (CAS 9011‑18‑1) is a versatile high‑molecular‑weight polyanion, essential in molecular biology (e.g., hybridisation), biochemistry (e.g., protein/lipoprotein precipitation), virology, inflammation modeling, and nanomedicine construction
- White to off-white crystalline powder
- BSE/TSE Free
- Loss on drying: ≤10%
- Free Sulfate: ≤0,5 %
- pH (1%, Water) @ 25C: 5,0 to 7,5
- Chloride: ≤1000 ppm
- Sulfur Content: 17,0 to 20,0%
Synonymes: Dextran sulphate sodium from Dextran
- Numéro CAS:9011-18-1
- Poids moléculaire:>500,000 g/mol
Spécifications
Spécifications
- Type de conditionnement:Bottle
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



