Dextran sulphate
Bottle
Catalog # RC-044
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Dextran sulfate sodium (CAS 9011‑18‑1) is a versatile high‑molecular‑weight polyanion, essential in molecular biology (e.g., hybridisation), biochemistry (e.g., protein/lipoprotein precipitation), virology, inflammation modeling, and nanomedicine construction
- White to off-white crystalline powder
- BSE/TSE Free
- Loss on drying: ≤10%
- Free Sulfate: ≤0,5 %
- pH (1%, Water) @ 25C: 5,0 to 7,5
- Chloride: ≤1000 ppm
- Sulfur Content: 17,0 to 20,0%
Synonyms: Dextran sulphate sodium from Dextran
- CAS Number:9011-18-1
- Molecular Weight:>500,000 g/mol
Specifications
Specifications
- Type de conditionnement:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



