Microtubes à centrifuger, Axygen®
0,5 ml, incolore, avec fileté, sans bouchon, non graduée, fond conique, non stériles, 500 tubes/pack
N° de catalogue AXYGST-45
Fournisseur: Corning
Aller à
Product Details & Documents
Screw cap tubes made of PP feature a cap that provides a tight secure seal.
- Certified RNase-/DNase-free, nonpyrogenic
- Will withstand up to 20000×g
- Screw caps to be ordered seperately
Screw cap tubes are available in a variety of colors with a conical or self-standing bottom.
Documents
Spécifications
- Emballage:500 tubes/pack
- Type:avec fileté, sans bouchon
- Version:non graduée, fond conique, non stériles
- Capacité:0,5 ml
- Couleur:incolore
- Seq:02