Potassium hydroxyde 25% (p/v) en solution aqueuse
Glass bottle, 1 L
CAS No.: 1310-58-3 Molecular Formula: HKO Formula Weight: 56.11 Physical Form: Liquid UN No.: 1814 MDL No.: MFCD00003553
: Potash caustic, Caustic potash
- :1310-58-3
- :KOH
- :MFCD00003553
- :56.11 g/mol
- :Température ambiante
- :215-181-3
- :1814
- :8,II
- Type de conditionnement:Glass bottle
- Cdt:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



