Plongeurs de tubes
Plongeurs de tubes
Tubing sinkers, fits tubing with inner Ø 5,5 - 6,2 mm
Catalog # BELAF329030102
Plongeurs de tubes
Catalog # BELAF329030102
Plongeurs de tubes
Plongeurs de tubes
Tubing sinkers, fits tubing with inner Ø 5,5 - 6,2 mm
Price
SFr. 76.50
Log in for availability
1 * 1 ST
Product Details & Documents
These Teflon® sinkers help to keep tubing at the bottom of the container.
  • Ideal for use with pumps and dispensers
  • Hose barb ends for simple tubing connection
  • Solvent resistant and autoclavable
Documents
Specifications
  • Désignation:
    Tubing sinkers, fits tubing with inner Ø 5,5 - 6,2 mm
  • Seq:
    02